Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200-1:2,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:150 | - |
Immunocytochemistry (ICC/IF) |
1:100 | - |
Product Specifications | |
---|---|
Species Reactivity |
Dog, Human, Mouse |
Host/Isotype |
Mouse / IgG2a |
Class |
Monoclonal |
Type |
Antibody |
Clone |
OTI1D7 |
Immunogen |
Full length human recombinant protein of human UBE2G2 produced in HEK293T cell. |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS with 1% BSA, 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein shares 100% sequence identity with the mouse counterpart. This gene is ubiquitously expressed, with high expression seen in adult muscle. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2 ubiquitin-conjugating enzyme G2; Ubiquitin carrier protein G2; ubiquitin conjugating enzyme 7; ubiquitin conjugating enzyme E2G 2; ubiquitin conjugating enzyme G2; ubiquitin-conjugating enzyme 7 homolog; Ubiquitin-conjugating enzyme E2 G2; ubiquitin-conjugating enzyme E2G 2 (homologous to yeast UBC7); ubiquitin-conjugating enzyme E2G 2 (UBC7 homolog, yeast); Ubiquitin-protein ligase G2
Gene Aliases: 1110003O05Rik; D10Xrf369; UBC7; Ubc7p; UBE2G2
UniProt ID: (Human) P60604, (Mouse) P60605
Entrez Gene ID: (Human) 7327, (Dog) 611581, (Mouse) 22213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support