Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ADEEAGGTER MEISAELPQT PQRLASWWDQ QVDFYTAFLH HLAQLVPEIY FAEMDPDLEK QEESVQMSIF TPLEWYLFGE DPDICLEKLK HSGAFQLCG
ELK1 is a component of the ternary complex that binds the serum response element (SRE) and mediates gene activity in response to serum and growth factors. ELK1 is phosphorylated by MAP kinase pathways at a cluster of S/T motifs at its C-terminus. Phosphorylation at these sites, particularly Ser383, is critical for transcriptional activation by ELK1. ELK1 appears to be a direct target of activated MAP kinase. Biochemical studies indicate that ELK1 is a good substrate for MAP kinase, the kinetics of ELK1 phosphorylation and activation correlate with MAP kinase activity, and interfering mutants of MAP kinase block ELK1 activation in vivo. ELK1 is a nuclear target for the ras-raf-MAPK signaling cascade. Alternatively spliced transcript variants encoding the same protein have been found for ELK1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase UBR1; E3a ligase; MGC142065; MGC142067; N-recognin-1; RING-type E3 ubiquitin transferase UBR1; ubiquitin ligase E3 alpha-I; ubiquitin-protein ligase E3-alpha; Ubiquitin-protein ligase E3-alpha-1; Ubiquitin-protein ligase E3-alpha-I
Gene Aliases: JBS; UBR1
UniProt ID: (Human) Q8IWV7
Entrez Gene ID: (Human) 197131
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support