Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic FGF repressed, Zic binding protein; basic FGF repressed, zinc binding protein; Basic FGF-repressed Zic-binding protein; bFGF-repressed Zic-binding protein; cytochrome B protein synthesis 3 homolog; FLJ10850; mbFZb; Ubiquinol-cytochrome c reductase complex chaperone CBP3 homolog; ubiquinol-cytochrome c reductase complex chaperone, CBP3 homolog; Ubiquinol-cytochrome-c reductase complex assembly factor 1
Gene Aliases: Bfzb; BZFB; C20orf44; UQCC; UQCC1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support