Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP24 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 24; Ubiquitin carboxyl-terminal hydrolase 24; ubiquitin specific protease 24; Ubiquitin thioesterase 24; ubiquitin thiolesterase 24; ubiquitin-specific processing protease 24; Ubiquitin-specific-processing protease 24
Gene Aliases: 2700066K03Rik; 2810030C21Rik; AI414051; B130021E18; C79851; KIAA1057; USP24
UniProt ID: (Human) Q9UPU5, (Mouse) B1AY13
Entrez Gene ID: (Human) 23358, (Mouse) 329908
Molecular Function: basic helix-loop-helix transcription factor cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support