Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Deubiquitinating enzyme exhibiting a preference towards 'Lys-63'-linked Ubiquitin chains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 38; HP43.8KD; Ubiquitin carboxyl-terminal hydrolase 38; ubiquitin specific protease 38; Ubiquitin thioesterase 38; ubiquitin thiolesterase 38; Ubiquitin-specific-processing protease 38
Gene Aliases: HP43.8KD; KIAA1891; USP38
UniProt ID: (Human) Q8NB14
Entrez Gene ID: (Human) 84640, (Dog) 476071
Molecular Function: cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support