Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji.
Predicted reactivity: Mouse (90%), Rat (90%), Sheep (96%), Bovine (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLIC1 is an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and is found in lesions associated with Alzheimer's and Parkinson's disease.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPLIC-1; PLIC-1; Protein linking IAP with cytoskeleton 1; testicular tissue protein Li 219; Ubiquilin-1
Gene Aliases: 1110046H03Rik; 1810030E05Rik; AU019746; C77538; D13Ertd372e; DA41; DSK2; PLIC-1; PLIC1; UBQLN1; UBQN; XDRP1
UniProt ID: (Human) Q9H3R4, (Mouse) Q8R317
Entrez Gene ID: (Human) 29979, (Mouse) 56085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support