Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Anti-VDAC/Porin Antibody is directed against human VDAC1/Porin1 protein. A BLAST analysis was used to suggest that this antibody would react with VDAC1/Porin1 from a wide range or organisms, including avian, mammalian, aquatic and reptilian sources based on 100% homology for the immunogen sequence. Cross reactivity will occur with all forms of VDACs including VDAC1, VDAC2 (4 isoforms) and VDAC3 (2 isoforms). Such broad reactivity makes this antibody useful as an excellent loading control (mitochondrial).
Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC constitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BR1-VDAC; Brain-derived voltage-dependent anion channel 1; hVDAC1; MGC111064; Mitochondrial Porin; mVDAC1; mVDAC5; Outer mitochondrial membrane protein porin 1; Plasmalemmal porin; Porin 31HL; Porin 31HM; PORIN-31-HL; rVDAC1; VD; VDAC-1; VDAC-5; Voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5
Gene Aliases: AL033343; PORIN; VDAC; VDAC-1; VDAC1; Vdac5
UniProt ID: (Bovine) P45879, (Human) P21796, (Pig) Q9MZ16, (Mouse) Q60932, (Rat) Q9Z2L0
Entrez Gene ID: (Bovine) 282119, (Human) 7416, (Pig) 397010, (Goat) 102173290, (Mouse) 22333, (Rat) 83529
Molecular Function: voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support