Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
p193-4 reacts with an internal epitope (amino acids 491-494, HPGE) of the minor vault protein (p193 or VPARP), which is overexpressed in various human non-P-glycoprotein MDR tumor cell lines, accordingly to an increase in the number of vault particles. p193-4 was raised against an E. coli lysate transformed with the pET28a(+) expression vector containing amino acids 408-611 of the p193 cDNA.
This gene encodes poly(ADP-ribosyl)transferase-like 1 protein, which is capable of catalyzing a poly(ADP-ribosyl)ation reaction. This protein has a catalytic domain which is homologous to that of poly (ADP-ribosyl) transferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly (ADP-ribosyl) transferase. Since this protein is not capable of binding DNA directly, its transferase activity may be activated by other factors such as protein-protein interaction mediated by the extensive carboxyl terminus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 193 kDa; 193 kDa vault protein; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 1; ADP-ribosyltransferase diphtheria toxin-like 4; H5 proline-rich; I-alpha-I-related; PARP-related; PARP-related/IalphaI-related H5/proline-rich; poly [ADP-ribose] polymerase 4; poly(ADP-ribose) synthetase; poly(ADP-ribosyl)transferase-like 1; unnamed protein product; vault poly(ADP-ribose) polymerase; vault protein, 193-kDa; von Willebrand factor A domain containing 5C
Gene Aliases: ADPRTL1; ARTD4; p193; PARP-4; PARPL; PH5P; VAULT3; VPARP; VWA5C
UniProt ID: (Human) Q9UKK3
Entrez Gene ID: (Human) 143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support