Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:300-5,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Tag |
Host/Isotype |
Mouse / IgG |
Class |
Monoclonal |
Type |
Antibody |
Clone |
1000000 |
Immunogen |
KLH conjugated VSV-G tag |
Conjugate |
HRP
|
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
TBS with 1% BSA, 50% glycerol |
Contains |
0.03% ProClin 300 |
Storage conditions |
-20°C or -80°C if preferred |
Vesicular stomatitis virus (VSV), an enveloped RNA virus from the Rhabdoviridae family, is released from the plasma membrane of host cells by a process called budding. The glycoprotein (VSV G) contains a domain in its extracellular membrane-proximal stem that appears to be needed for efficient VSV budding: Sequence: YTDIEMNRLGK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: vesicular stomatitis virus; VSIVgp3; VSV epitope tag; VSV tag; VSV-G epitope tag; vsvg; VSVG epitope tag; VSVG tag; YTDIEMNRLGK
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support