Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: APGGGQHGLF GPGAGGLHHA HSDAQGHLLF PGLPEQHGPH GSQNVLNGQM RLGLPGEVFG RSEQYRQVAS PRTDPYSAAQ LHNQYGPMNM N
ZIC2 (zinc finger protein of the cerebellum 2) is a transcriptional repressor and may regulate tissue specific expression of dopamine receptor D1. In mouse, knockout of Zic1 leads to aplasia of cerebellum and skeletal abnormalities. Mutation in Zic2 and Zic3 in mouse and/or human cause holoprosencephaly and heterotaxis, respectively. Holoprosencephaly is the most common structural anomaly of the human brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zic family member 2 (odd-paired homolog, Drosophila); Zinc finger protein of the cerebellum 2; Zinc finger protein ZIC 2
Gene Aliases: HPE5; ZIC2
UniProt ID: (Human) O95409
Entrez Gene ID: (Human) 7546
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support