Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Human Relaxin-1 quantitates human Relaxin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Relaxin-1.
Relaxins are known endocrine and autocrine/paracrine hormones, belonging to the insulin gene superfamily. In the human there are three non-allelic relaxin genes, RLN1, RLN2 and RLN3. RLN1 and RLN2 share high sequence homology. This encoded protein is synthesized as a single-chain polypeptide but the active form consists of an A chain and a B chain linked by disulfide bonds; however, their exact cleavage sites have not been described. Relaxin is produced by the ovary, and targets the mammalian reproductive system to ripen the cervix, elongate the pubic symphysis and inhibit uterine contraction. It may have additional roles in enhancing sperm motility, regulating blood pressure, controlling heart rate and releasing oxytocin and vasopressin. This gene has multiple polyadenylation sites. There are multiple alternatively spliced transcript variants described for this gene but their full length nature is not known yet.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : bA12D24.3.1, bA12D24.3.2, H1, H1RLX, RLN1, RLXH1
Gene ID : (Human) 6013
Gene symbol : RLN1
Protein Aliases : preprorelaxin H1, Prorelaxin H1, Relaxin A chain, Relaxin B chain
UniProt ID (Human) P04808
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support