Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Rat IP-10 (CXCL10) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant Rat IP-10 (CXCL10) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay Rat IP-10 (CXCL10) in approximately 1,000 ELISA plate wells.
This PeproTech ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant proteins in a sandwich ELISA format. This ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells. The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : Cxcl10, Inp10, IP-10, Mob1, Scyb10
Gene ID : (Rat) 245920
Gene symbol : Cxcl10
Protein Aliases : 10 kDa interferon gamma-induced protein, C-X-C motif chemokine 10, Gamma-IP10, interferon-inducible cytokine IP-10, Interferon-inducible protein 10, Protein Mob-1, small inducible cytokine B subfamily (Cys-X-Cys) member 10, small inducible cytokine B subfamily (Cys-X-Cys), member 10, Small-inducible cytokine B10
UniProt ID (Rat) P48973
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support