Search
Basic Sandwich ELISA Protocols |
Enzyme-linked immunosorbent assays (ELISAs) are plate-based assays used to detect and quantify a specific protein in complex mixture. In a sandwich ELISA, highly specific antibodies immobilize the target protein (antigen) to the plate and indirectly detects the presence of the target protein. This type of capture assay is called a sandwich assay because the analyte is bound between two primary antibodies, each detecting a different epitope of the antigen—the capture antibody and the detection antibody.
Find protocols below for a standard sandwich ELISA using a 96-well plate for the detection techniques—colorimetric (chromogenic), chemiluminescent, and fluorescent detection.
Search available antibody pairs and ELISA kits Browse available ELISA reagents Build your own ELISA
This protocol represents an example colorimetric sandwich ELISA using direct detection with an HRP-conjugated antibody or indirect detection with a biotinylated antibody and streptavidin-HRP.
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
If using alkaline phosphatase (AP) instead of HRP for the enzyme conjugate, an AP-specific substrate must be used. Substitute TMB substrate solution in step 15 with p-nitrophenyl phosphate (PNPP) (1-Step PNPP Substrate Solution). Incubate at room temperature for 15–30 minutes. Substitute the stop solution with 50 µL of 2 N NaOH to stop the reaction. Measure absorbance of each well at 405 nm.
When immobilizing the antigen-containing sample directly to the plate, there is no need for a capture antibody. Different concentrations of the sample should be prepared in coating buffer and identical volumes added directly to the plate. The rest of the protocol should be performed as previously described using a detection antibody and enzyme conjugate plus substrate.
When using a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, there will be slightly less amplification of enzyme signal compared to using a biotinylated detection antibody with streptavidin-HRP. Therefore, it may be necessary to use a slightly higher concentration of secondary antibody-enzyme conjugate than one would normally use for a streptavidin-enzyme conjugate.
This protocol represents an example of a chemiluminescent sandwich ELISA using direct detection with an HRP-conjugated antibody or indirect detection with a biotinylated antibody and streptavidin-HRP. A luminol-based substrate is used for detection.
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
When substituting horseradish peroxidase (HRP) with alkaline phosphatase (AP) for the enzyme conjugate, it is necessary to use an AP-specific substrate. Replace the TMB substrate solution in step 15 with p-nitrophenyl phosphate (PNPP) (1-Step PNPP Substrate Solution). Incubate the reaction mixture at room temperature for 15 to 30 minutes. To terminate the reaction, add 50 µL of 2 N NaOH as the stop solution. Subsequently, measure the absorbance of each well at 405 nm.
When directly immobilizing the antigen-containing sample onto the plate, a capture antibody is not required. Prepare varying concentrations of the sample in coating buffer and add equal volumes directly to the plate. Follow the remainder of the protocol as previously outlined, utilizing a detection antibody along with an enzyme conjugate and a substrate.
When employing a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, the amplification of the enzyme signal will be somewhat reduced compared to using a biotinylated detection antibody with streptavidin-HRP. Consequently, it may be required to use a slightly higher concentration of the secondary antibody-enzyme conjugate than typically used for a streptavidin-enzyme conjugate.
This protocol represents an example of a fluorescent sandwich ELISA using direct detection with an HRP-conjugated antibody or indirect detection with a biotinylated antibody and streptavidin-HRP. A fluorogenic peroxidase substrate is used for detection.
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
With a fluorophore-conjugated detection antibody, no enzyme conjugate or substrate is required. The plate fluorescence units can be measured directly after step 11. The working concentration of labeled antibody or protein is typically 2–4 μg/mL.
For direct immobilization of the antigen-containing sample onto the plate, a capture antibody is not needed. Prepare the sample in varying concentrations using the coating buffer and dispense equal volumes directly onto the plate. Continue with the rest of the protocol as previously detailed, applying a detection antibody along with an enzyme conjugate and substrate.
When utilizing a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, the enzyme signal amplification will be lower compared to the use of a biotinylated detection antibody with streptavidin-HRP. Therefore, you might need to increase the concentration of the secondary antibody-enzyme conjugate slightly more than what is usually required for a streptavidin-enzyme conjugate.
The following tables provide recommended ranges for different ELISA components. Concentrations are guidelines only; for best results, optimize each component individually.
Recommended starting concentration ranges for coating and detection antibodies for ELISA optimization. The use of non-purified antibodies will work but may result in higher background. It is generally recommended to use affinity-purified antibodies for optimal signal-to-noise ratio.
| Source | Coating antibody | Detection antibody |
| Polyclonal serum | 5–15 μg/mL | 1–10 μg/mL |
| Crude ascites | 5–15 μg/mL | 1–10 μg/mL |
| Affinity-purified polyclonal antibody | 1–12 μg/mL | 0.5–5 μg/mL |
| Affinity-purified monoclonal antibody | 1–12 μg/mL | 0.5–5 μg/mL |
Recommended detection antibody concentration for ELISA in different systems. Check the user guide for the substrate as it may recommend a more defined concentration range for the enzyme conjugate.
| Enzyme | System | Concentration |
|---|---|---|
| HRP | Colorimetric | 20–200 ng/mL |
| Chemifluorescent | 25–50 ng/mL | |
| Chemiluminescent | 10–100 ng/mL | |
| AP | Colorimetric | 100–200 ng/mL |
| Chemiluminescent | 40–200 ng/mL |
When a non-biotinylated detection antibody is used after an enzyme-labeled secondary antibody, the resulting enzyme signal amplification is slightly reduced compared to the use of a biotinylated detection antibody with streptavidin-HRP. As a result, it may be necessary to use a slightly higher concentration of the secondary antibody-enzyme conjugate than typically used for a streptavidin-enzyme conjugate.
For a complete set of ELISA reagents, Invitrogen ELISA Buffer Kit includes 2 Coating Buffers (pH 7.4 and pH 9.4), 5X Assay Buffer (blocking reagent and diluent), 25X Wash Buffer, Stabilized TMB, and Stop Solution. Pair this kit with Matched Antibody Pair kits that contain matched antibody pairs, standards and Streptavidin-HRP conjugate.
For Research Use Only. Not for use in diagnostic procedures.