Sealing Tape for 96-Well Plates
Thermo Scientific™
Sealing Tape for 96-Well Plates
Thermo Scientific Microplate Sealing Tape is a package of pre-cut, pressure-sensitive sheets for sealing the surface of standard 96-well orRead more
Have Questions?
Catalog NumberQuantity
15036100 sheets
Catalog number 15036
Price (USD)
159.00
Each
Quantity:
100 sheets
Price (USD)
159.00
Each
Thermo Scientific Microplate Sealing Tape is a package of pre-cut, pressure-sensitive sheets for sealing the surface of standard 96-well or 384-well microplates during incubation steps.
Features of Microplate Sealing Tape:
• Pressure-sensitive—position the tape over plate and press to seal; easy to remove
• Separate sheets—each sheet of tape is supplied on its own easy-to-peal backing (liner)
• Durable and inert—film thickness provides support and has a thin layer of acrylic adhesive
Use the disposable sheets to seal and reseal a microplate two or three times for incubation steps in an ELISA or protein assay procedure. Seal plates quickly and easily to allow mixing or prevent evaporation during assay incubation steps.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeFilm, Seal, or Applicator
Quantity100 sheets
AdhesivePressure Sensitive
ColorClear
MaterialPolyester
No. of Wells96/384
Plate Compatibility96/384-well Plate
SterilityNon-sterile
Unit SizeEach