Product Image
Thermo Scientific™

HRP-Conjugated Streptavidin

HRP-Conjugated Streptavidin consists of streptavidin protein that is covalently conjugated to horseradish peroxidase (HRP) enzyme (RZ > 3.0). Streptavidin bindsRead more
Have Questions?
Catalog NumberQuantity
N1000.25 mL
Catalog number N100
Price (USD)
155.65
Online exclusive
173.00
Save 17.35
Each
Quantity:
0.25 mL
Request bulk or custom format
Price (USD)
155.65
Online exclusive
173.00
Save 17.35
Each
HRP-Conjugated Streptavidin consists of streptavidin protein that is covalently conjugated to horseradish peroxidase (HRP) enzyme (RZ > 3.0). Streptavidin binds to biotin and the conjugated HRP provides enzyme activity for detection using an appropriate substrate system. This particular product has been used primarily in sandwich ELISA applications to provide consistent measurement of biotinylated detection antibodies.

Features of HRP-Conjugated Streptavidin :

• Product Number: N100
• Size: 0.25 mL
• Concentration: 1.25 mg/mL
• Formulation: Provided in a conjugate stabilizer containing 0.1% ProClin Compound as a preservative.
• Storage: Shipped on cold packs. Store at 2-8°C upon receipt. Protect from exposure to light. Stable until expiration date if stored under these conditions.
• Application: ELISA: to be used with biotin-labeled detection antibodies. Recommended dilution range is from 1:4,000 to 1:20,000. The optimal dilution must be determined empirically for each specific application. Prepare dilutions immediately before use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1.25 mg/mL
Label or DyeHRP (Horseradish Peroxidase)
Product TypeStreptavidin Conjugate
Quantity0.25 mL
Target MoleculeBiotin
ConjugateHRP
FormLiquid
Unit SizeEach
Contents & Storage
Store between 2°C and 8°C.

Frequently asked questions (FAQs)

In which solvent is the HRP-Conjugated Streptavidin (Cat. No. N100) dissolved?

The HRP-Conjugated Streptavidin (Cat. No. N100) is dissolved in an aqueous solution of 0.99% BSA and 0.1% ProCelin preservative.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.