Pierce™ 96-Well Polystyrene Plates, White Opaque
Pierce™ 96-Well Polystyrene Plates, White Opaque
Pierce™ 96-Well Polystyrene Plates, White Opaque
Thermo Scientific™
Pierce™ 96-Well Polystyrene Plates, White Opaque
Thermo Scientific Pierce White Opaque 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics.Read more
Have Questions?
Catalog NumberQuantity
1504225 plates
Catalog number 15042
Price (USD)
161.65
Online exclusive
188.00
Save 26.35
Each
Quantity:
25 plates
Price (USD)
161.65
Online exclusive
188.00
Save 26.35
Each
Thermo Scientific Pierce White Opaque 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics. These microplates have a corner notch for use in standard plate readers.
Features of White Opaque 96-Well Microplates:
• White opaque plates for fluorescent or chemiluminescent assays
• Treated to provide protein coating capacity
• High antibody-binding surface improves the overall performance in immunoassay applications
• Plates are certified for performance, having consistently low coefficients of variation among wells
Plate Properties:
• Antibody binding capacity: High, 400 ng IgG/cm2 (1 cm2 = 100 μL volume)
• Binding interaction: hydrophobic-ionic
• Well-to-well coefficient of variation: CV <10%
• Well shape (bottom): Flat
• Maximum well volume: 360 μL
Related Products
Pierce™ 96-Well Polystyrene Plates, Corner Notch
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyAdsorption, Hydrophilic
DescriptionPierce 96-Well Polystyrene Plates, White Opaque
Detection MethodColorimetric
Packaging25 Plate
Plate BlockingNone
Product TypeWell Plate
Quantity25 plates
ColorWhite
MaterialPolystyrene
No. of Wells96
Product LinePierce
SterilityNon-sterile
Surface TreatmentPolystyrene
Well ShapeFlat Bottom (F-well)
Unit SizeEach
Frequently asked questions (FAQs)
What kind of 96-well plate do I need for my fluorescent assay?
Select a microplate that is compatible with your instrument. Refer to the manufacturer's manual for types of microplates that are recommended for use with your instrument.
Various types of plastic may autofluoresce. Select a microplate that is rated for fluorescence-based assays and exhibits little or no autofluorescence in the emission range of the reagent you wish to use.
Clear plates are routinely used for fluorescence-based assays, but opaque-walled microplates reduce bleed-through between wells. If you are using live cells, you may need to consider special substrate treatments for the bottom of the wells, such as poly-L-lysine.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.