Thermo Scientific Chemicals

L-Aspartic acid, 99%, low metals content, Thermo Scientific Chemicals

Catalog number: 043317.22
100 g, Each
Thermo Scientific Chemicals

L-Aspartic acid, 99%, low metals content, Thermo Scientific Chemicals

Catalog number: 043317.22
100 g, Each
Quantity
Catalog number: 043317.22
also known as 043317-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
56-84-8
IUPAC Name
2-aminobutanedioic acid
Molecular Formula
C4H7NO4
InChI Key
CKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILES
NC(CC(O)=O)C(O)=O
Assay (unspecified)
> 98.5%
Form
Powder
Loss on Drying
< 0.2 %
Appearance (Color)
White

Description

L-Aspartic acid is used as a component of parenteral and enteral nutrition and as a pharmaceutical ingredient. it is used for cell culture and in manufacturing processes. It is widely utilized for mineral supplementation in the salt form.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text