DRI™ pH-Detect™ Specimen Validity Test Calibrators
Thermo Scientific™
DRI™ pH-Detect™ Specimen Validity Test Calibrators
Combat the use of additives and adulterants to mask illicit drugs in urine samples with Thermo Scientific™ DRI™ pH-Detect Specimen Validity Test. Thermo Scientific provides two calibrators to calibrate the tests.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpH
1002833.0 & 11.0
10024403TS
also known as 10024403
4.0 & 11.0
Catalog number 100283
Price (USD)
-
pH:
3.0 & 11.0
Combat the use of additives and adulterants to mask illicit drugs in urine samples with Thermo Scientific™ DRI™ pH-Detect Specimen Validity Test. Thermo Scientific provides two calibrators to calibrate the tests. Liquid and ready-to-use, the tests can be performed on a variety of general chemistry analyzers.
The use of additives and adulterants to mask illicit drugs in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution. The adulteration tests provide a complete and automated way to test for sample integrity and adulteration.
  • Complete and automated way to test for sample integrity and adulteration
  • Associated controls offered in convenient sizes
  • The 4.0 & 11.0 pH calibrators support new SAMHSA guidelines (effective October 1, 2017)
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSpecimen Validity Test Calibrator Set
Quantity2 x 25 mL (1 x 25 mL pH 3.0 and 1 x 25 mL pH 11.0)
TypeCalibrator
Detectable AnalytespH
Sample TypeUrine
pH3.0 & 11.0
Unit SizeEach
Contents & Storage
2°C to 8°C