Thermo Scientific™

ImmunoCAP™ Mite Allergens

Catalog number: 14410701
Thermo Scientific™

ImmunoCAP™ Mite Allergens

Catalog number: 14410701
Quantify specific IgE antibodies with ImmunoCAP™ Mite Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
 
Catalog Number
14410701
Unit Size
Each
Common Name
House dust mite
Scientific Name
Dermatophagoides pteronyssinus
Code
d1
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameDermatophagoides pteronyssinus
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity16 Tests
Coded1
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
House dust mite
Scientific Name
Dermatophagoides farinae
Code
d2
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameDermatophagoides farinae
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity16 Tests
Coded2
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
House dust mite
Scientific Name
Dermatophagoides microceras
Code
d3
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameDermatophagoides microceras
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity16 Tests
Coded3
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Storage mite
Scientific Name
Lepidoglyphus destructor
Code
d71
Quantity
10 Tests
Price (USD)
Full specifications
Scientific NameLepidoglyphus destructor
Regulatory StatusFDA Cleared
Code TypeMite
Common NameStorage mite
Quantity10 Tests
Coded71
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Storage mite
Scientific Name
Acarus siro
Code
d70
Quantity
10 Tests
Price (USD)
Full specifications
Scientific NameAcarus siro
Regulatory StatusFDA Cleared
Code TypeMite
Common NameStorage mite
Quantity10 Tests
Coded70
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Storage mite
Scientific Name
Tyrophagus putrescentiae
Code
d72
Quantity
10 Tests
Price (USD)
Full specifications
Scientific NameTyrophagus putrescentiae
Regulatory StatusFDA Cleared
Code TypeMite
Common NameStorage mite
Quantity10 Tests
Coded72
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
House dust mite
Scientific Name
Euroglyphus maynei
Code
d74
Quantity
10 Tests
Price (USD)
Full specifications
Scientific NameEuroglyphus maynei
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity10 Tests
Coded74
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Storage mite
Scientific Name
Glycyphagus domesticus
Code
d73
Quantity
10 Tests
Price (USD)
Full specifications
Scientific NameGlycyphagus domesticus
Regulatory StatusFDA Cleared
Code TypeMite
Common NameStorage mite
Quantity10 Tests
Coded73
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
House dust mite
Scientific Name
Blomia tropicalis
Code
d201
Quantity
10 Tests
Price (USD)
Full specifications
Scientific NameBlomia tropicalis
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity10 Tests
Coded201
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeCommon NameScientific NameCodeQuantityPrice (USD)
14410701Full specifications
EachHouse dust miteDermatophagoides pteronyssinusd116 TestsRequest A Quote
Scientific NameDermatophagoides pteronyssinus
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity16 Tests
Coded1
Unit SizeEach
14410801Full specifications
EachHouse dust miteDermatophagoides farinaed216 TestsRequest A Quote
Scientific NameDermatophagoides farinae
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity16 Tests
Coded2
Unit SizeEach
14424101Full specifications
EachHouse dust miteDermatophagoides microcerasd316 TestsRequest A Quote
Scientific NameDermatophagoides microceras
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity16 Tests
Coded3
Unit SizeEach
14430101Full specifications
EachStorage miteLepidoglyphus destructord7110 TestsRequest A Quote
Scientific NameLepidoglyphus destructor
Regulatory StatusFDA Cleared
Code TypeMite
Common NameStorage mite
Quantity10 Tests
Coded71
Unit SizeEach
14430001Full specifications
EachStorage miteAcarus sirod7010 TestsRequest A Quote
Scientific NameAcarus siro
Regulatory StatusFDA Cleared
Code TypeMite
Common NameStorage mite
Quantity10 Tests
Coded70
Unit SizeEach
14430201Full specifications
EachStorage miteTyrophagus putrescentiaed7210 TestsRequest A Quote
Scientific NameTyrophagus putrescentiae
Regulatory StatusFDA Cleared
Code TypeMite
Common NameStorage mite
Quantity10 Tests
Coded72
Unit SizeEach
14434201Full specifications
EachHouse dust miteEuroglyphus mayneid7410 TestsRequest A Quote
Scientific NameEuroglyphus maynei
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity10 Tests
Coded74
Unit SizeEach
14439301Full specifications
EachStorage miteGlycyphagus domesticusd7310 TestsRequest A Quote
Scientific NameGlycyphagus domesticus
Regulatory StatusFDA Cleared
Code TypeMite
Common NameStorage mite
Quantity10 Tests
Coded73
Unit SizeEach
14489601Full specifications
EachHouse dust miteBlomia tropicalisd20110 TestsRequest A Quote
Scientific NameBlomia tropicalis
Regulatory StatusFDA Cleared
Code TypeMite
Common NameHouse dust mite
Quantity10 Tests
Coded201
Unit SizeEach
Showing 9 of 9
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text