Thermo Scientific Chemicals

L(+)-Norleucine, ≥99%, Thermo Scientific Chemicals

Catalog number: 226040050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

L(+)-Norleucine, ≥99%, Thermo Scientific Chemicals

Catalog number: 226040050
5 g, Glass Bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
327-57-1
IUPAC Name
(2S)-2-aminohexanoic acid
Molecular Formula
C6H13NO2
InChI Key
LRQKBLKVPFOOQJ-YFKPBYRVSA-N
SMILES
CCCC[C@H](N)C(O)=O
Appearance (Form)
Crystalline powder
Titration with HClO4
>=99.0 %
Other amino acids
(TLC) Not detectable
Appearance (Color)
White to almost white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text