Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Dispense contents without tipping or shaking with Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles. Wide mouth allows faster, easier filling. Bottle and tubulation are molded in one piece for uniform service and precision flow without sputtering. The tip can be cut back to increase flow. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. Naturally flexible LDPE material is chemically compatible with a wide range of laboratory reagents and provides an ergonomic soft squeeze to help reduce hand fatigue.
Closure Material | Polypropylene |
Closure Size | 24 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 48.3 mm |
Height (English) | 4.7 in. |
Height (Metric) | 119.4 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 4 oz., 125 mL |
Capacity (Metric) | 125 mL |
Capacity (English) | 4 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 48 |
No. per Pack | 6 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Pack of 6 |
Closure Material | Polypropylene |
Closure Size | 24 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 48.3 mm |
Height (English) | 4.7 in. |
Height (Metric) | 119.4 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 4 oz., 125 mL |
Capacity (Metric) | 125 mL |
Capacity (English) | 4 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 48 |
No. per Pack | 6 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Case of 48 |
Closure Material | Polypropylene |
Closure Size | 24 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 62.5 mm |
Height (English) | 5.42 in. |
Height (Metric) | 137.7 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 8 oz., 250 mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 36 |
No. per Pack | 4 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Case of 36 |
Closure Material | Polypropylene |
Closure Size | 24 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 62.5 mm |
Height (English) | 5.42 in. |
Height (Metric) | 137.7 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 8 oz., 250 mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 36 |
No. per Pack | 4 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Pack of 4 |
Closure Material | Polypropylene |
Closure Size | 38 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 76.7 mm |
Height (English) | 6.81 in. |
Height (Metric) | 173 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Case of 24 |
Closure Material | Polypropylene |
Closure Size | 38 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 76.7 mm |
Height (English) | 6.81 in. |
Height (Metric) | 173 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Pack of 4 |
Closure Material | Polypropylene |
Closure Size | 38 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 84.8 mm |
Height (English) | 8.05 in. |
Height (Metric) | 204.5 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 24 oz., 750 mL |
Capacity (Metric) | 750 mL |
Capacity (English) | 24 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Case of 24 |
Closure Material | Polypropylene |
Closure Size | 38 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 84.8 mm |
Height (English) | 8.05 in. |
Height (Metric) | 204.5 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 24 oz., 750 mL |
Capacity (Metric) | 750 mL |
Capacity (English) | 24 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Pack of 4 |
Closure Material | Polypropylene |
Closure Size | 43 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 96.5 mm |
Height (English) | 8.25 in. |
Height (Metric) | 209.6 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 32 oz., 1000 mL |
Capacity (Metric) | 1000 mL |
Capacity (English) | 32 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 12 |
No. per Pack | 2 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Case of 12 |
Closure Material | Polypropylene |
Closure Size | 43 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 96.5 mm |
Height (English) | 8.25 in. |
Height (Metric) | 209.6 mm |
Through the Closure Dispensing | No |
Product Type | Wash Bottle |
Capacity | 32 oz., 1000 mL |
Capacity (Metric) | 1000 mL |
Capacity (English) | 32 oz. |
Color | Translucent |
Description | LDPE bottle/tubulation |
Material | LDPE |
No. per Case | 12 |
No. per Pack | 2 |
Closure Included | Yes |
Graduated | No |
Mouth | Wide |
Shape | Cylindrical |
Unit Size | Pack of 2 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | dimClosureSize | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2402-0125PK also known as 2402-0125 | Pack of 6 | 125 mL | Price: 54.90 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2402-0125 | Case of 48 | 125 mL | Price: 372.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2402-0250 | Case of 36 | 250 mL | Price: 316.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2402-0250PK also known as 2402-0250 | Pack of 4 | 250 mL | Price: 41.50 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2402-0500 | Case of 24 | 500 mL | Price: 259.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2402-0500PK also known as 2402-0500 | Pack of 4 | 500 mL | Price: 51.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2402-0750 | Case of 24 | 750 mL | Price: 317.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2402-0750PK also known as 2402-0750 | Pack of 4 | 750 mL | Price: 62.70 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2402-1000 | Case of 12 | 1000 mL | Price: 188.00 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2402-1000PK also known as 2402-1000 | Pack of 2 | 1000 mL | Price: 37.20 Your price: | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Includes:
Polypropylene screw closure
.