Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | ART 200U, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 200 μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 200U |
Length (English) | 2.06 in. |
Length (Metric) | 5.2cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Ergonomic |
For Use With (Equipment) | Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Pack of 960 |
Description | ART 1000 ERGO-F, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 1000 μL |
Filtered | Filtered |
Sterility | Sterile |
Model | ART 1000 ERGO-F |
Length (English) | 2.78 in. |
Length (Metric) | 7.061cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Ergonomic |
For Use With (Equipment) | Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Each |
Description | ART 200U, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 200 μL |
Filtered | Filtered |
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Model | ART 200U |
Length (English) | 2.06 in. |
Length (Metric) | 5.2cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Ergonomic |
For Use With (Equipment) | Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Case of 4800 |
Description | ART 50U, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 50 μL |
Filtered | Filtered |
Sterility | Sterile |
Model | ART 50U |
Length (English) | 2.06 in. |
Length (Metric) | 5.2cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Ergonomic |
For Use With (Equipment) | Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Case of 4800 |
Description | ART 50U, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 50 μL |
Filtered | Filtered |
Sterility | Sterile |
Model | ART 50U |
Length (English) | 2.06 in. |
Length (Metric) | 5.2cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Ergonomic |
For Use With (Equipment) | Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Pack of 960 |
Description | ART 1000, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 1000 μL |
Filtered | Filtered |
Sterility | Sterile |
Model | ART 1000 |
Length (English) | 3.13 in. |
Length (Metric) | 7.9cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Ergonomic |
For Use With (Equipment) | Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Case of 3200 |
Description | ART 1000, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 1000 μL |
Filtered | Filtered |
Sterility | Sterile |
Model | ART 1000 |
Length (English) | 3.13 in. |
Length (Metric) | 7.9cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Ergonomic |
For Use With (Equipment) | Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Pack of 800 |
Description | ART 10 REACH, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 10 μL |
Filtered | Filtered |
Sterility | Sterile |
Model | ART 10 REACH |
Length (English) | 1.71 in. |
Length (Metric) | 4.33cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Extended Length |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Case of 4800 |
Description | ART 10 REACH, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 10 μL |
Filtered | Filtered |
Sterility | Sterile |
Model | ART 10 REACH |
Length (English) | 1.71 in. |
Length (Metric) | 4.33cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Extended Length |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Pack of 960 |
Description | ART 20E, Filtered, Sterile, Lift-off Lid Rack |
Volume (Metric) | 0.5 to 20 μL |
Filtered | Filtered |
Sterility | Sterile |
Model | ART 20E |
Length (English) | 1.78 in. |
Length (Metric) | 4.521cm |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Tip Style | Micro Point |
For Use With (Equipment) | Eppendorf™ Pipettor |
Product Line | ART |
Fit for Purpose | Sterile & PCR Ready |
Unit Size | Case of 4800 |
Catalog Number | Specifications | Unit Size | Description | Volume (Metric) | Tip Style | Fit for Purpose | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2770PK also known as 2770 | Pack of 960 | ART 200U, Filtered, Sterile, Lift-off Lid Rack | 200 μL | Ergonomic | Sterile & PCR Ready | Price: 179.50 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2380 | Each | ART 1000 ERGO-F, Filtered, Sterile, Lift-off Lid Rack | 1000 μL | Ergonomic | Sterile & PCR Ready | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2770 | Case of 4800 | ART 200U, Filtered, Sterile, Lift-off Lid Rack | 200 μL | Ergonomic | Sterile & PCR Ready | Price: 868.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2780 | Case of 4800 | ART 50U, Filtered, Sterile, Lift-off Lid Rack | 50 μL | Ergonomic | Sterile & PCR Ready | Price: 883.00 Online price: 618.10 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2780PK also known as 2780 | Pack of 960 | ART 50U, Filtered, Sterile, Lift-off Lid Rack | 50 μL | Ergonomic | Sterile & PCR Ready | Price: 181.70 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2279 | Case of 3200 | ART 1000, Filtered, Sterile, Lift-off Lid Rack | 1000 μL | Ergonomic | Sterile & PCR Ready | Price: 718.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2279PK also known as 2279 | Pack of 800 | ART 1000, Filtered, Sterile, Lift-off Lid Rack | 1000 μL | Ergonomic | Sterile & PCR Ready | Price: 184.20 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2140 | Case of 4800 | ART 10 REACH, Filtered, Sterile, Lift-off Lid Rack | 10 μL | Extended Length | Sterile & PCR Ready | Price: 928.00 Online price: 649.60 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2140PK also known as 2140 | Pack of 960 | ART 10 REACH, Filtered, Sterile, Lift-off Lid Rack | 10 μL | Extended Length | Sterile & PCR Ready | Price: 212.60 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2149 | Case of 4800 | ART 20E, Filtered, Sterile, Lift-off Lid Rack | 0.5 to 20 μL | Micro Point | Sterile & PCR Ready | Price: 878.00 Online price: 745.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features:
Designed to Aid in Pipetting for a Variety of Applications:
Extended length: to prevent contamination by keeping the pipette shaft from entering the sample vessel
Gel loading: for loading acrylamide or agarose gels
Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials
Solvent safe: for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes