Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product Type | Cup, Microcentrifuge |
Product Line | Slide-A-Lyzer™ |
MWCO | 2.0 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device, 0.1 mL |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Quantity | 250 Devices |
Unit Size | 250 devices |
Product Type | Cup, Microcentrifuge |
Product Line | Slide-A-Lyzer™ |
MWCO | 2.0 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device, 0.1 mL |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Quantity | 50 Devices |
Unit Size | 50 devices |
Catalog Number | Specifications | Unit Size | Sample Volume (Metric) | Quantity | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
69553 | 250 devices | 0.1 mL | 250 Devices | 666.65 Online price 748.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
69580 | 50 devices | 0.1 mL | 50 Devices | 215.65 Online price 242.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The 0.1mL MINI Dialysis Devices are designed to be capped (caps included) and inserted into standard 1.5mL microcentrifuge tubes. Alternatively, as many as 25 MINI Devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple inside a large beaker. Using the 0.1mL MINI Device, 100μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes.