VersaTREK™ Myco Susceptibility Kit
VersaTREK™ Myco Susceptibility Kit
VersaTREK™ Myco Susceptibility Kit
Thermo Scientific™
VersaTREK™ Myco Susceptibility Kit
Perform comprehensive Mtb susceptibility testing with a kit that includes three FDA-cleared antituberculosis drugs: rifampin, isoniazid and ethambutol.
Have Questions?
Catalog NumberPackaging TypeProduct Type
7115-6080 Tests/KitMicrobiology Media
Catalog number 7115-60
Price (USD)
-
Packaging Type:
80 Tests/Kit
Product Type:
Microbiology Media
Perform comprehensive Mtb susceptibility testing with three FDA-cleared antituberculosis drugs using the Thermo Scientific™ VersaTREK™ Myco Susceptibility Kit. The kit includes rifampin, isoniazid and ethambutol.
The VersaTREK System combines a liquid culture medium (VersaTREK Myco media), a growth supplement (Myco GS), and, for potentially contaminated species, an antibiotic supplement (Myco AS or Myco PVNA) with a detection system that automatically incubates and continuously monitors culture bottles inoculated with specimens possibly containing mycobacteria.
The VersaTREK Myco Susceptibility Kit consists of three primary antituberculosis drugs: Rifampin (1μg/mL), Isoniazid (0.4μg/mL and 0.1μg/mL), and Ethambutol (8μg/mL and 5μg/mL).
Mycobacteria susceptibility testing also requires VersaTREK Myco Growth Supplement (Myco GS, Part No. 7112-42) and VersaTREK Myco Antibiotic Supplement (Myco AS, Part No. 7114-42).
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Antimicrobial AgentsRifampin, Isoniazid, and Ethambutol
DescriptionVersaTREK Myco Susceptibility Kit
No. per Pack80 Tests/Kit
Packaging Type80 Tests/Kit
Quantity2 x 25 mL Rifampin, 2 x 25 mL Isoniazid, 3 x 25 mL Ethambutol
Specimen TypeAll Samples (e.g. Sputum, Blood, Tissue, Urine, etc.)
Product TypeMicrobiology Media
SpeciesMycobacterium tuberculosis
Target Organism ClassMycobacteria
Test TypeMicrobial Detection, Antimicrobial Susceptibility
Unit SizeEach
Contents & Storage
2°C to 8°C