Search Thermo Fisher Scientific
Type | Validator Kit |
Description | iCAP 6000 Validator Kit - IG Toolkit including Aqueous test solutions |
For Use With (Equipment) | iCAP 6000 Series ICP-OES System |
Unit Size | - |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
842313050701 | - | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The iCAP 6000 Validator Kit for aqueous sample matrices includes the following:
Compatibility
Compatible with all iCAP 6000 Series ICP-OES systems which are operating the most current version of iTEVA software (backwards compatible to iCAP Issue 8.2 software)