Search Thermo Fisher Scientific
Type | Autosampler |
Description | CETAC EXR-8 Autosampler |
For Use With (Equipment) | iCAP 7000 Series ICP-OES System |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
842347004003 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
CETAC ASX-260 Autosampler
Offers a 180 sample capacity for low-moderate sample throughput, compatible with:
CETAC ASX-520 Autosampler
Accommodates up to 360 samples for moderate-high sample throughput, compatible with:
CETAC ASX-520 Q Dual Rinse Autosampler
Only for use with the iCAP Q Series ICP-MS system when the Autosampler Housing is deployedCETAC XLR-8 Autosampler
Enables the unattended analysis of up to 720 samples, for laboratories looking to push the boundaries of unattended analysis, compatible with:
CETAC ASX-1400 Autosampler
Incorporates automated liquid stirring to ensure sample homogeneity, for samples containing particulates in suspension such as wear metals applications, compatible with;