Thermo Scientific Chemicals

D-(-)-Arabinose, 99%

Catalog number: A10357.09
10 g, Each
Thermo Scientific Chemicals

D-(-)-Arabinose, 99%

Catalog number: A10357.09
10 g, Each
Quantity
Catalog number: A10357.09
Price (USD)
Quantity
-

Chemical Identifiers

CAS
10323-20-3
IUPAC Name
oxane-2,3,4,5-tetrol
Molecular Formula
C5H10O5
InChI Key
SRBFZHDQGSBBOR-UHFFFAOYNA-N
SMILES
OC1COC(O)C(O)C1O
Appearance (Color)
White to cream
Form
Powder
Assay (Silylated GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
152.0-161.0?C

Description

D-(-)-Arabinose is used in culture media for some bacteria. It serves as a component of biopolymers such as hemicellulose and pectin. In the synthetic biology laboratory, it is employed as a reversible switch for protein expression under the Pbad promoter (i.e. part of plasmids) in E. coli. Further, it acts as an inhibitor of the enzyme glucose dehydrogenase. In addition to this, it is used as a pharmaceutical intermediate and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Arabinose is used in culture media for some bacteria. It serves as a component of biopolymers such as hemicellulose and pectin. In the synthetic biology laboratory, it is employed as a reversible switch for protein expression under the Pbad promoter (i.e. part of plasmids) in E. coli. Further, it acts as an inhibitor of the enzyme glucose dehydrogenase. In addition to this, it is used as a pharmaceutical intermediate and food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text