Thermo Scientific Chemicals

L-(+)-Arabinose, 99%

Catalog number: A11921.18
50 g, Each
Thermo Scientific Chemicals

L-(+)-Arabinose, 99%

Catalog number: A11921.18
50 g, Each
Quantity
Catalog number: A11921.18
also known as A11921-18
Price (USD)
Price: 84.40
Online price: 71.65
Your price:
Quantity
-

Chemical Identifiers

CAS
87-72-9
IUPAC Name
(3S,4R,5R)-oxane-2,3,4,5-tetrol
Molecular Formula
C5H10O5
InChI Key
SRBFZHDQGSBBOR-ZRMNMSDTSA-N
SMILES
O[C@@H]1COC(O)[C@@H](O)[C@@H]1O
Appearance (Color)
White to pale cream
Assay (HPLC)
≥98.5%
Optical Rotation
+104? ± 2? (c=10 in Water, 24h)
Form
Powder
Identification (FTIR)
Conforms

Description

L-(+)-Arabinose is used to identify and differentiate and characterize pentose sugar isomerase. It is also used as the carbon source in microbial culture. It is involved in the bioproduction of L-ribose. It finds application in the production of savory reaction flavors. Its derivatives are used in the development of antiviral agents such as nucleoside. It acts as a base component of hemicellulose and pectin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Arabinose is used to identify and differentiate and characterize pentose sugar isomerase. It is also used as the carbon source in microbial culture. It is involved in the bioproduction of L-ribose. It finds application in the production of savory reaction flavors. Its derivatives are used in the development of antiviral agents such as nucleoside. It acts as a base component of hemicellulose and pectin.

Solubility
Soluble in water and glycerol. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text