Thermo Scientific Chemicals

Methyl alpha-D-glucopyranoside, 98%, Thermo Scientific Chemicals

Catalog number: A12484.22
100 g, Each
Thermo Scientific Chemicals

Methyl alpha-D-glucopyranoside, 98%, Thermo Scientific Chemicals

Catalog number: A12484.22
100 g, Each
Quantity
Catalog number: A12484.22
also known as A12484-22
Price (USD)
Price: 38.10
Online price: 36.65
Your price:
Quantity
-

Chemical Identifiers

CAS
97-30-3
IUPAC Name
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular Formula
C7H14O6
InChI Key
HOVAGTYPODGVJG-UHFFFAOYNA-N
SMILES
COC1OC(CO)C(O)C(O)C1O
Identification (FTIR)
Conforms
Melting Point
165.0-170.0?C
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤0.5%
Optical Rotation
+159? ± 3? (c=10 in water)

Description

Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.

Solubility
Soluble in water. Insoluble in ether

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text