Thermo Scientific Chemicals

D-(+)-Cellobiose, 98+%

Catalog number: A14553.14
25 g, Each
Thermo Scientific Chemicals

D-(+)-Cellobiose, 98+%

Catalog number: A14553.14
25 g, Each
Quantity
Catalog number: A14553.14
also known as A14553-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
528-50-7
IUPAC Name
2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
GUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILES
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
Form
Crystals or powder or crystalline powder
Optical Rotation
+34? ?1? (c=10, in Water for 15 hours)
Appearance (Color)
White
Identification (FTIR)
Conforms
Solution Test
5% w/v in Water: Clear

Description

D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.

Solubility
Soluble in water (1 g/8ml). Insoluble in ether and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text