Thermo Scientific Chemicals

Diethylcarbamyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A15701.14
25 g, Each
Thermo Scientific Chemicals

Diethylcarbamyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A15701.14
25 g, Each
Quantity
Catalog number: A15701.14
also known as A15701-14
Price (USD)
Price: 22.30
Online price: 17.65
Your price:
Quantity
-

Chemical Identifiers

CAS
88-10-8
IUPAC Name
N,N-diethylcarbamoyl chloride
Molecular Formula
C5H10ClNO
InChI Key
OFCCYDUUBNUJIB-UHFFFAOYSA-N
SMILES
CCN(CC)C(Cl)=O
Identification (FTIR)
Conforms (U.K. specification)
Form
Liquid
Assay from Suppliers CofA
≥97.5% (GC, U.S. specification)
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5% (U.K. specification)

Description

It is used in the production of pharma ingredients. It is a reagent in the synthesis of S-Methyl-N,N-diethylthiocarbamate Sulfoxide, which is an oxygenated metabolite of Disulfiram (Antabuse) that is capable of in vitro inactivation of liver mitochondrial aldehyde dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the production of pharma ingredients. It is a reagent in the synthesis of S-Methyl-N,N-diethylthiocarbamate Sulfoxide, which is an oxygenated metabolite of Disulfiram (Antabuse) that is capable of in vitro inactivation of liver mitochondrial aldehyde dehydrogenase.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text