Thermo Scientific Chemicals

L-Methionine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A15952.14
25 g, Each
Thermo Scientific Chemicals

L-Methionine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A15952.14
25 g, Each
Quantity
Catalog number: A15952.14
also known as A15952-14
Price (USD)
Price: 110.00
Online price: 93.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2491-18-1
IUPAC Name
hydrogen methyl 2-amino-4-(methylsulfanyl)butanoate chloride
Molecular Formula
C6H14ClNO2S
InChI Key
MEVUPUNLVKELNV-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC(=O)C(N)CCSC
Assay from Supplier's CofA
≥98.5%
Optical Rotation
+26.0° ± 1.5° (c=1, water)
Appearance (Color)
White to pale cream
Form
Powder or crystals or crystalline powder

Description

L-Methionine methyl ester hydrochloride is used as a starting material for the synthesis of a protected glycine derivative, a versatile asymmetric building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Methionine methyl ester hydrochloride is used as a starting material for the synthesis of a protected glycine derivative, a versatile asymmetric building block.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text