IonCode™ Barcode Adapters 1-384 Kit
Ion Torrent™

IonCode™ Barcode Adapters 1-384 Kit

The IonCode Barcode Adapters 1-384 Kit provides a set of 384 unique barcode adapters specifically designed for optimal performance with the Ion GeneStudio S5, Ion PGM, and Ion Proton semiconductor sequencers.
Have Questions?
Catalog NumberQuantity
A297513840 Reactions
Catalog number A29751
Price (USD)
14,420.00
Each
Quantity:
3840 Reactions
Price (USD)
14,420.00
Each

The IonCode Barcode Adapters 1-384 Kit provides a set of 384 unique barcode adapters specifically designed for optimal performance with the Ion GeneStudio S5, Ion PGM, and Ion Proton semiconductor sequencers. When used in combination with Ion Barcode Adapters 385-768 (Cat. No. A36546) and Ion AmpliSeq Library Kit PLUS or Ion AmpliSeq Library Kit 2.0, this kit enables users to pool up to 768 amplicon libraries and then conduct multiplexed sequencing analysis, simplifying the Ion semiconductor sequencing workflow for a wide range of applications, including targeted enrichment.

Features of the IonCode Barcode Adapters 1-384 Kit include:

  • Enables multiplexing of numerous amplicon library samples on a single sequencing chip with the use of robust molecular barcodes
  • Both sequence- and flow-optimized for equal representation of all barcodes in a pool and more economical multiplexed sequencing runs
  • Robust error correction built in by design for added confidence in sample identification

Multiplexing with barcode adapters enables higher throughput
Multiplexing with barcoded libraries leads to more cost effective runs by permitting up to 768 samples per run, significantly decreasing the cost and handling requirements of sequencing. Each barcode was individually tested to minimize representation bias.

Sequence- and flow-optimized for increased performance and efficiency
Barcode adapters contain sequences that are optimized to provide equal representation of all barcodes in a pool and require minimal number of flows to interrogate, resulting in more economical multiplexed sequencing runs.

Robust error correction built in by design
Built-in error correction prevents the possibility of incorrect sample assignment.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Detection and Identification of Animal Disease
For Use With (Equipment)Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
LibrariesAmplicon Library
No. of Reactions3840
Product LineIonCode
Product TypeBarcode Adapter Kit
Quantity3840 Reactions
Sample TypeDNA
Shipping ConditionDry Ice
Workflow StepLibrary Barcoding
FormatKit
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
Contents & Storage
4 x 96 well plates, 20 μL per well

Store in freezer (-5°C to -30°C).

Frequently asked questions (FAQs)

What do I need to do to prep the barcode adapters in the IonCode Barcode Adapters 1-96 Kit (Cat. No. A29747)?

When handling barcode adapters, be careful to avoid cross-contamination by changing gloves frequently and by carefully removing the plate seal.
- Pulse vortex the IonCode Barcode Adapters plate several times.
- Centrifuge the IonCode Barcode Adapters plate for 1 minute at 1000 x g to collect contents.
- Carefully remove, then discard the plate seal.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many reactions are possible with the IonCode Barcode Adapters 1-96 Kit (Cat. No. A29747)?

The IonCode Barcode Adapters 1-96 Kit consists of 1 x 96-well plate, with 20 µL of each barcode per well. The plate consists of 960 reactions in total with 10 reactions for each of the 96 barcodes.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

In what format are the barcode adapters in the IonCode Barcode Adapters 1-96 Kit (Cat. No. A29747)?

The IonCode Barcode Adapters 1-96 Kit consists of 1 x 96-well plate, with 20 µL of each barcode per well. The barcodes are pre-diluted, ready-to-use. The 96-well plate format allows multichannel pipetting for more efficient work.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What are the shipping and storage conditions for IonCode Barcode Adapters 1-96 Kit (Cat. No. A29747)?

The IonCode Barcode Adapters 1-96 Kit is shipped on dry ice and we recommend storing at -30 degrees C to -5 degrees C in an upright position.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many samples from the Oncomine Comprehensive Assay Plus, manual library preparation can I run on the Ion 550 Chip?

We recommend sequencing up to 5 DNA or RNA samples (4 samples and a non-template control (NTC)) on a single Ion 550 Chip.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.