Oncomine™ BCR IGH SR Assay, RNA
Ion Torrent™

Oncomine™ BCR IGH SR Assay, RNA

The Oncomine BCR IGH-SR Assay, RNA, is a robust, targeted next-generation sequencing (NGS) assay designed for hematology-oncology and minimal residualRead more
Have Questions?
Catalog NumberNo. of Reactions
A4548424
Catalog number A45484
Price (USD)
7,700.00
Each
No. of Reactions:
24
Price (USD)
7,700.00
Each

The Oncomine BCR IGH-SR Assay, RNA, is a robust, targeted next-generation sequencing (NGS) assay designed for hematology-oncology and minimal residual disease (MRD) monitoring research, pharmacodynamics and biomarker analysis of immunotherapy, as well as infectious and autoimmune disease research. The assay kit includes a single pool of multiplex PCR primers, total RNA-to-cDNA synthesis kit and Ion AmpliSeq library reagents.

The assay accurately identifies and measures clonal expansion of B lymphocytes using total RNA from bone marrow, whole blood, peripheral blood leukocytes (PBLs), peripheral blood mononuclear cells (PBMCs), and fresh-frozen and formalin-fixed paraffin-embedded (FFPE) research samples. The assay identifies unique B-cell clones through targeting of the highly diverse complementarity-determining region 3 (CDR3) of the B-cell receptor IGH chain from mRNA template via AmpliSeq multiplex framework 3 and joining gene primers. The nucleotide sequence of the IGH CDR3 region serves as a natural barcode to enable clone tracking for MRD clinical research and measurements of B-cell clonal expansion and diversity for pharmacodynamics and vaccine biomarker research. IGH CDR3 region amino acid motifs may reveal signatures of B-cell responses to defined antigens for use as future markers of autoimmune or infectious disease. RNA input improves detection of changes in plasmablast and plasma cell populations following immune challenge or administration of immunomodulatory agents. For DNA samples, please see Cat. No. A45483.

Benefits of the Oncomine BCR IGH SR Assay, RNA, include:

  • High sensitivity and exceptionally low limit of detection for rare clone detection and tracking in research samples via dual-barcode indexing and high template capture efficiency of AmpliSeq multiplex PCR
  • Compatibility with an array of research sample types, including fresh-frozen and FFPE tissue, bone marrow, whole blood, PBLs, and PBMCs
  • Assay primers design to amplify all variable and joining gene alleles in the gold-standard IMGT database
  • Ability to generate large libraries with minimal input requirement, facilitating detection of rare clones at a frequency of 10-6 from a single library preparation
  • Efficient workflow using the Ion Chef templating system and Ion GeneStudio S5 sequencing system, enabling library-to-results time within 48 hours
  • Flexible input requirements ranging from 25 ng to 2 μg
  • Streamlined and user-friendly informatics solution with automated clonotyping, clonal lineage analysis, reporting of key repertoire features, and multi-sample analysis capability to enable longitudinal tracking in research samples
  • Ultra-low substitution error rate minimizing artifacts arising from sequencing errors, enabling highly accurate assessment of clonal heterogeneity in malignant B-cell research samples of interest

The Oncomine BCR IGH-SR Assay, RNA, uses multiplex PCR primers to generate 110-bp amplicons that can be sequenced on all chip types supported by the Ion GeneStudio S5 sequencing systems, allowing you to pick the best multiplexing configuration for your unique sample batching needs and throughput requirements.

Learn more about the assay

Note: Information about data analysis using Ion Reporter Software v5.12 can be found in the User Guide below.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chip CompatibilityIon 520, Ion 530, Ion 540, Ion 550
For Use With (Application)Immune Repertoire Research
For Use With (Equipment)Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
LibrariesAmplicon
No. of Reactions24
No. of SamplesUp to 24 Samples
Nucleic Acid TypeRNA
Product LineOncomine
Product TypeAssay
Sample TypeBone Marrow, Whole Blood, PBMC, Fresh/Frozen Tissue, FFPE Tissue
Shipping ConditionApproved for shipment on Wet or Dry Ice
ThroughputChip-dependent Multiplexing
Workflow StepPanel and Library
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.

Frequently asked questions (FAQs)

How many libraries can be prepped with each Oncomine BCR IGH SR Assay, RNA?

The assay provides sufficient reagents for rapid preparation of 24 sample libraries.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is included in the Oncomine BCR IGH SR Assay, RNA?

The Oncomine BCR IGH SR Assay, RNA consists of the Oncomine BCR IGH-SR Panel, the Ion AmpliSeq Library Kit Plus (Cat. No. 4488990), and the Ion Torrent NGS Reverse Transcription Kit (Cat. No. A45003). Sufficient reagents are provided for the rapid preparation of 24 sample libraries.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is the input requirement for the Oncomine BCR IGH SR Assay, RNA?

The assay input requirement ranges from 25 ng to 2 µg DNase-treated total RNA. For RNA samples with a concentration of <6.25 ng/µL, we recommend using the DNAclear Purification Kit (Cat. No. AM1756) to concentrate the sample after performing reverse transcription. We recommend using ≥ 50 ng DNase-treated total RNA depending on sample type, ranging from 50-2,000 ng (see "Saturation analysis of peripheral blood leukocytes (PBL) - Oncomine BCR IGH Assay" on page 159 and "Saturation analysis of tissue - Oncomine BCR IGH Assay" on page 161 of the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017438_OncomineTCRBetaAssay_UG.pdf).

When using degraded samples (FFPE) or samples with unknown B or T-cell content, perform sample qualification and quantitation to determine the minimum sample input that is required. For more information, see Appendix B, "Sample qualification and quantitation" on page 130 of the user guide.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What types of samples is the Oncomine BCR IGH SR Assay, RNA compatible with?

This assay is compatible with isolated B cells, whole blood, peripheral blood leukocytes (PBLs), peripheral blood mononuclear cells (PBMCs), and fresh frozen tissue samples, including FFPE and bone marrow samples.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is the Oncomine BCR IGH SR Assay, RNA?

The Oncomine BCR IGH SR Assay, RNA (Cat. No. A45484) is a highly sensitive RNA-based NGS assay that enables the characterization of the immunoglobulin heavy chain complementarity-determining region 3 (CDR3) sequences. The assay accurately measures B cell repertoire diversity and clonal expansion in a wide variety of sample types.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.