Gibco™
Xenon™ SingleShot Electroporation Chamber
Catalog number: A53444
Gibco™
Xenon™ SingleShot Electroporation Chamber
Catalog number: A53444
Catalog Number
A53444
Unit Size
6 chambers
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A534446 chambers
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Gibco Xenon SingleShot Electroporation Chamber holds a volume of 1 mL for electroporation for cell therapy research with the Xenon Electroporation System. The single-use electroporation chamber is made of Tritan™ MX711 copolyester with gold-plated electrodes and is sterilized by gamma irradiation. It can process 2 × 107 to 1 × 108 T cells in a 1-mL sample volume and is available in a pack of six individually packaged chambers.
Promising electroporation conditions that have been optimized in the Neon Transfection System can be scaled to the Xenon Electroporation System without sacrificing performance.
View an interactive 3D tour of the CTS Xenon instrument ›
Specifications
Capacity (Metric)
1 mL
Quantity
6 chambers
For Use With (Equipment)
Gibco Xenon Electroporation System