Thermo Scientific™

AMS-4 Beta Air Monitor

Catalog number: AMS4OPT13
Thermo Scientific™

AMS-4 Beta Air Monitor

Catalog number: AMS4OPT13

Provide an early warning to workers exposed to potential airborne releases of beta-emitting particulates when using Thermo Scientific™ AMS-4 Beta Air Monitor. Lightweight and robust design accommodates both fixed and portable use applications.

 
Catalog Number
AMS4OPT13
Unit Size
Each
Description
Monitoring and calibration software
Price (USD)
Full specifications
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
DescriptionMonitoring and calibration software
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeSystem Monitoring and Calibration Software with Cables for Beta Air Monitor
Power Consumption1.5 A
Voltage220 V
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Display unit without sampling head
Price (USD)
Full specifications
Alarm/Fail RelaysSPST, 15 A at 30 VDC/120 VAC resistive
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
ConnectionsComputer: DB9, RS-232 standard, RS-485 optional
Data Log Buffer2000 entries of combined Concentration and Status
DescriptionDisplay unit without sampling head
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Detector TypeSealed proportional window
Flow Rate8.5 to 113 L/min.
Outputs0 to 5 VDC, five decade logarithmic
Radiation TypeBeta
Display TypeTwo rows x 20 characters high visibility vacuum fluorescent, percentage of Alarm 40-element LED bar graph
Product TypeBeta Air Monitor Display Unit
Keypad18 keys, full numeric plus function keys
Status IndicatorFront panel lights display READY and MALFUNCTION conditions, red alarm strobe light, sonalert
Window Density2 to 3 mg/cm²
Width (English)11 in.
Width (Metric)27.9 cm
Height (English)8.75 in.
Height (Metric)22.2 cm
Depth (English)2.8 in.
Depth (Metric)32.5 cm
Diameter (Metric) Detector Window50.8 mm
Weight (English)7.5 lb.
Weight (Metric)3.4 kg
Electrical Requirements100 to 240 V 50/60 Hz
Voltage100 V to 240 V
Wattage120 W
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Radial entry sampling head
Price (USD)
Full specifications
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
DescriptionRadial entry sampling head
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Efficiency8.5% 60Co, 17% 90Sr/90Y (nominal).
Product TypeRadial Entry Sampling Head for Beta Air Monitor
Width (English)5.6 in.
Width (Metric)14.2 cm
Height (English)7.3 in.
Height (Metric)18.5 cm
Depth (English)9.8 in.
Depth (Metric)24.9 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RS-485 serial interface
Price (USD)
Full specifications
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
DescriptionRS-485 serial interface
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeRS-485 Serial Interface for Beta Air Monitor
Unit SizeEach
Catalog Number
Unit Size
Each
Description
25 ft. cable/tubing for OPT14 and OPT8
Price (USD)
Full specifications
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
Description25 ft. cable/tubing for OPT14 and OPT8
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeExtension Cable/Tubing for Beta Air Monitor
Depth (English)25 ft.
Depth (Metric)7.6 m
Unit SizeEach
Catalog Number
Unit Size
Each
Description
50 ft. cable/tubing for OPT14 and OPT8
Price (USD)
Full specifications
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
Description50 ft. cable/tubing for OPT14 and OPT8
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeExtension Cable/Tubing for Beta Air Monitor
Depth (English)50 ft.
Depth (Metric)15.2 m
Unit SizeEach
Catalog Number
Unit Size
Each
Description
In-line entry sampling head
Price (USD)
Full specifications
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
DescriptionIn-line entry sampling head
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
EfficiencyRadial Sampling Head: 8.5% 60Co, 17% 90Sr/90Y (nominal); Inline Sampling Head: 6.4% 85Kr, 4.4% 133Xe (nominal); Noble Gas Sampling Head: 5.75% 60Co, 12% 90Sr/90Y (nominal)
Product TypeIn-line Entry Sampling Head for Beta Air Monitor
Width (English)5.6 in.
Width (Metric)14.2 cm
Height (English)7.3 in.
Height (Metric)18.5 cm
Depth (English)11.6 in.
Depth (Metric)29.5 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Noble gas sampling head
Price (USD)
Full specifications
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
DescriptionNoble gas sampling head
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
EfficiencyRadial Sampling Head: 8.5% 60Co, 17% 90Sr/90Y (nominal); Inline Sampling Head: 6.4% 85Kr, 4.4% 133Xe (nominal); Noble Gas Sampling Head: 5.75% 60Co, 12% 90Sr/90Y (nominal)
Product TypeNoble Gas Sampling Head for Beta Air Monitor
Width (English)9.3 in.
Width (Metric)23.6 cm
Height (English)6.4 in.
Height (Metric)16.3 cm
Depth (English)9.5 in.
Depth (Metric)24.1 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
15 ft. extension cable/tubing for OPT14 and OPT8 only
Price (USD)
Full specifications
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
Description15 ft. extension cable/tubing for OPT14 and OPT8 only
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeExtension Cable/Tubing for Beta Air Monitor
Depth (English)15 ft.
Depth (Metric)4.6 m
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Depot service warranty for AMS-4
Price (USD)
Full specifications
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
DescriptionDepot service warranty for AMS-4
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeService Warranty for Beta Air Monitor
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
AMS4OPT13Full specifications
EachMonitoring and calibration softwareRequest A Quote
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
DescriptionMonitoring and calibration software
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeSystem Monitoring and Calibration Software with Cables for Beta Air Monitor
Power Consumption1.5 A
Voltage220 V
Unit SizeEach
AMS4Full specifications
EachDisplay unit without sampling headRequest A Quote
Alarm/Fail RelaysSPST, 15 A at 30 VDC/120 VAC resistive
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
ConnectionsComputer: DB9, RS-232 standard, RS-485 optional
Data Log Buffer2000 entries of combined Concentration and Status
DescriptionDisplay unit without sampling head
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Detector TypeSealed proportional window
Flow Rate8.5 to 113 L/min.
Outputs0 to 5 VDC, five decade logarithmic
Radiation TypeBeta
Display TypeTwo rows x 20 characters high visibility vacuum fluorescent, percentage of Alarm 40-element LED bar graph
Product TypeBeta Air Monitor Display Unit
Keypad18 keys, full numeric plus function keys
Status IndicatorFront panel lights display READY and MALFUNCTION conditions, red alarm strobe light, sonalert
Window Density2 to 3 mg/cm²
Width (English)11 in.
Width (Metric)27.9 cm
Height (English)8.75 in.
Height (Metric)22.2 cm
Depth (English)2.8 in.
Depth (Metric)32.5 cm
Diameter (Metric) Detector Window50.8 mm
Weight (English)7.5 lb.
Weight (Metric)3.4 kg
Electrical Requirements100 to 240 V 50/60 Hz
Voltage100 V to 240 V
Wattage120 W
Frequency50/60 Hz
Unit SizeEach
AMS4OPT14Full specifications
EachRadial entry sampling headRequest A Quote
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
DescriptionRadial entry sampling head
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Efficiency8.5% 60Co, 17% 90Sr/90Y (nominal).
Product TypeRadial Entry Sampling Head for Beta Air Monitor
Width (English)5.6 in.
Width (Metric)14.2 cm
Height (English)7.3 in.
Height (Metric)18.5 cm
Depth (English)9.8 in.
Depth (Metric)24.9 cm
Unit SizeEach
AMS4OPT4Full specifications
EachRS-485 serial interfaceRequest A Quote
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
DescriptionRS-485 serial interface
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeRS-485 Serial Interface for Beta Air Monitor
Unit SizeEach
AMS4OPT6BFull specifications
Each25 ft. cable/tubing for OPT14 and OPT8Request A Quote
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
Description25 ft. cable/tubing for OPT14 and OPT8
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeExtension Cable/Tubing for Beta Air Monitor
Depth (English)25 ft.
Depth (Metric)7.6 m
Unit SizeEach
AMS4OPT6CFull specifications
Each50 ft. cable/tubing for OPT14 and OPT8Request A Quote
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
Description50 ft. cable/tubing for OPT14 and OPT8
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeExtension Cable/Tubing for Beta Air Monitor
Depth (English)50 ft.
Depth (Metric)15.2 m
Unit SizeEach
AMS4OPT7Full specifications
EachIn-line entry sampling headRequest A Quote
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
DescriptionIn-line entry sampling head
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
EfficiencyRadial Sampling Head: 8.5% 60Co, 17% 90Sr/90Y (nominal); Inline Sampling Head: 6.4% 85Kr, 4.4% 133Xe (nominal); Noble Gas Sampling Head: 5.75% 60Co, 12% 90Sr/90Y (nominal)
Product TypeIn-line Entry Sampling Head for Beta Air Monitor
Width (English)5.6 in.
Width (Metric)14.2 cm
Height (English)7.3 in.
Height (Metric)18.5 cm
Depth (English)11.6 in.
Depth (Metric)29.5 cm
Unit SizeEach
AMS4OPT8Full specifications
EachNoble gas sampling headRequest A Quote
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
DescriptionNoble gas sampling head
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
EfficiencyRadial Sampling Head: 8.5% 60Co, 17% 90Sr/90Y (nominal); Inline Sampling Head: 6.4% 85Kr, 4.4% 133Xe (nominal); Noble Gas Sampling Head: 5.75% 60Co, 12% 90Sr/90Y (nominal)
Product TypeNoble Gas Sampling Head for Beta Air Monitor
Width (English)9.3 in.
Width (Metric)23.6 cm
Height (English)6.4 in.
Height (Metric)16.3 cm
Depth (English)9.5 in.
Depth (Metric)24.1 cm
Unit SizeEach
AMS4OPT6AFull specifications
Each15 ft. extension cable/tubing for OPT14 and OPT8 onlyRequest A Quote
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
Description15 ft. extension cable/tubing for OPT14 and OPT8 only
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeExtension Cable/Tubing for Beta Air Monitor
Depth (English)15 ft.
Depth (Metric)4.6 m
Unit SizeEach
AMS4DS3Full specifications
EachDepot service warranty for AMS-4Request A Quote
Certifications/ComplianceMeets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
CompatibilityThermo Scientific ViewPoint Enterprise
DescriptionDepot service warranty for AMS-4
Alpha/Beta/Gamma MeasurementBeta Measurement
For Use With (Application)Beta Monitoring
Radiation TypeBeta
Product TypeService Warranty for Beta Air Monitor
Unit SizeEach
Showing 10 of 12Show more
Continuous air monitor (CAM) offering beta particulate, radioiodine, or noble gas detection.
  • Detachable sampling head
  • Reports both DAC and cpm
  • Real time gamma background subtraction
  • Stand alone or network configuration
  • Mass flow measurements
  • Fast and slow concentration alarms, cpm alarm, DAC-hour alarm
  • Computer calibration
  • Can also be used for stack monitoring of effluent releases using the in-line detector head
  • Keypad: 18 keys, full numeric plus function keys
  • Status Indicators: Front panel lights display READY and MALFUNCTION conditions, red alarm strobe light, sonalert
  • Printer: RS-232 serial DB9
  • Data Log Buffer: 2000 entries of combined Concentration and Status
  • Change information: Equivalent to 83 days of 1 hour detail, nearly 14 days of 10 mintes detail or over 24 hours of 1 minute detail
  • Alarm/Fail Relays: SPST, 15A at 30VDC/120VAC resistive
  • Analog Output: 0 to 5VDC output, five decade logarithmic

Compatible with:

Thermo Scientific ViewPoint Enterprise

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text