| AMS4OPT6A | Full specifications | Each | 15 ft. extension cable/tubing for OPT14 and OPT8 only | Request A Quote |
Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Description | 15 ft. extension cable/tubing for OPT14 and OPT8 only | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Radiation Type | Beta | Product Type | Extension Cable/Tubing for Beta Air Monitor | Depth (English) | 15 ft. | Depth (Metric) | 4.6 m | Unit Size | Each |
|
| AMS4 | Full specifications | Each | Display unit without sampling head | Request A Quote |
Alarm/Fail Relays | SPST, 15 A at 30 VDC/120 VAC resistive | Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Compatibility | Thermo Scientific ViewPoint Enterprise | Connections | Computer: DB9, RS-232 standard, RS-485 optional | Data Log Buffer | 2000 entries of combined Concentration and Status | Description | Display unit without sampling head | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Detector Type | Sealed proportional window | Flow Rate | 8.5 to 113 L/min. | Outputs | 0 to 5 VDC, five decade logarithmic | Radiation Type | Beta | Display Type | Two rows x 20 characters high visibility vacuum fluorescent, percentage of Alarm 40-element LED bar graph | Product Type | Beta Air Monitor Display Unit | Keypad | 18 keys, full numeric plus function keys | Status Indicator | Front panel lights display READY and MALFUNCTION conditions, red alarm strobe light, sonalert | Window Density | 2 to 3 mg/cm² | Width (English) | 11 in. | Width (Metric) | 27.9 cm | Height (English) | 8.75 in. | Height (Metric) | 22.2 cm | Depth (English) | 2.8 in. | Depth (Metric) | 32.5 cm | Diameter (Metric) Detector Window | 50.8 mm | Weight (English) | 7.5 lb. | Weight (Metric) | 3.4 kg | Electrical Requirements | 100 to 240 V 50/60 Hz | Voltage | 100 V to 240 V | Wattage | 120 W | Frequency | 50/60 Hz | Unit Size | Each |
|
| AMS4OPT13 | Full specifications | Each | Monitoring and calibration software | Request A Quote |
Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Compatibility | Thermo Scientific ViewPoint Enterprise | Description | Monitoring and calibration software | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Radiation Type | Beta | Product Type | System Monitoring and Calibration Software with Cables for Beta Air Monitor | Power Consumption | 1.5 A | Voltage | 220 V | Unit Size | Each |
|
| AMS4OPT14 | Full specifications | Each | Radial entry sampling head | Request A Quote |
Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Compatibility | Thermo Scientific ViewPoint Enterprise | Description | Radial entry sampling head | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Radiation Type | Beta | Efficiency | 8.5% 60Co, 17% 90Sr/90Y (nominal). | Product Type | Radial Entry Sampling Head for Beta Air Monitor | Width (English) | 5.6 in. | Width (Metric) | 14.2 cm | Height (English) | 7.3 in. | Height (Metric) | 18.5 cm | Depth (English) | 9.8 in. | Depth (Metric) | 24.9 cm | Unit Size | Each |
|
| AMS4OPT4 | Full specifications | Each | RS-485 serial interface | Request A Quote |
Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Compatibility | Thermo Scientific ViewPoint Enterprise | Description | RS-485 serial interface | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Radiation Type | Beta | Product Type | RS-485 Serial Interface for Beta Air Monitor | Unit Size | Each |
|
| AMS4OPT6B | Full specifications | Each | 25 ft. cable/tubing for OPT14 and OPT8 | Request A Quote |
Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Description | 25 ft. cable/tubing for OPT14 and OPT8 | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Radiation Type | Beta | Product Type | Extension Cable/Tubing for Beta Air Monitor | Depth (English) | 25 ft. | Depth (Metric) | 7.6 m | Unit Size | Each |
|
| AMS4OPT6C | Full specifications | Each | 50 ft. cable/tubing for OPT14 and OPT8 | Request A Quote |
Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Description | 50 ft. cable/tubing for OPT14 and OPT8 | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Radiation Type | Beta | Product Type | Extension Cable/Tubing for Beta Air Monitor | Depth (English) | 50 ft. | Depth (Metric) | 15.2 m | Unit Size | Each |
|
| AMS4OPT7 | Full specifications | Each | In-line entry sampling head | Request A Quote |
Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Description | In-line entry sampling head | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Radiation Type | Beta | Efficiency | Radial Sampling Head: 8.5% 60Co, 17% 90Sr/90Y (nominal); Inline Sampling Head: 6.4% 85Kr, 4.4% 133Xe (nominal); Noble Gas Sampling Head: 5.75% 60Co, 12% 90Sr/90Y (nominal) | Product Type | In-line Entry Sampling Head for Beta Air Monitor | Width (English) | 5.6 in. | Width (Metric) | 14.2 cm | Height (English) | 7.3 in. | Height (Metric) | 18.5 cm | Depth (English) | 11.6 in. | Depth (Metric) | 29.5 cm | Unit Size | Each |
|
| AMS4OPT8 | Full specifications | Each | Noble gas sampling head | Request A Quote |
Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Description | Noble gas sampling head | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Radiation Type | Beta | Efficiency | Radial Sampling Head: 8.5% 60Co, 17% 90Sr/90Y (nominal); Inline Sampling Head: 6.4% 85Kr, 4.4% 133Xe (nominal); Noble Gas Sampling Head: 5.75% 60Co, 12% 90Sr/90Y (nominal) | Product Type | Noble Gas Sampling Head for Beta Air Monitor | Width (English) | 9.3 in. | Width (Metric) | 23.6 cm | Height (English) | 6.4 in. | Height (Metric) | 16.3 cm | Depth (English) | 9.5 in. | Depth (Metric) | 24.1 cm | Unit Size | Each |
|
| AMS4DS3 | Full specifications | Each | Depot service warranty for AMS-4 | Request A Quote |
Certifications/Compliance | Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria | Compatibility | Thermo Scientific ViewPoint Enterprise | Description | Depot service warranty for AMS-4 | Alpha/Beta/Gamma Measurement | Beta Measurement | For Use With (Application) | Beta Monitoring | Radiation Type | Beta | Product Type | Service Warranty for Beta Air Monitor | Unit Size | Each |
|