Thermo Scientific™

Fluoro-Max Dyed Blue Aqueous Fluorescent Particles

Catalog number: B0200B
Thermo Scientific™

Fluoro-Max Dyed Blue Aqueous Fluorescent Particles

Catalog number: B0200B
Achieve improved sensitivity and detectability in analytical methods with Thermo Scientific™ Fluoro-Max Blue Aqueous Fluorescent Particles, internally dyed microsphere suspensions that feature bright, high-contrast colors.
 
Catalog Number
B0200B
Unit Size
Each
Quantity
60 mL
Diameter (Metric)
2 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity60 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean2 μm
Diameter (Metric) Nominal2 μm
Diameter (Metric)2 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
10 mL
Diameter (Metric)
1 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity10 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean1 μm
Diameter (Metric) Nominal1 μm
Diameter (Metric)1 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
60 mL
Diameter (Metric)
1 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity60 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean1 μm
Diameter (Metric) Nominal1 μm
Diameter (Metric)1 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
10 mL
Diameter (Metric)
2 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity10 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean2 μm
Diameter (Metric) Nominal2 μm
Diameter (Metric)2 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
15 mL
Diameter (Metric)
0.15 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity15 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<10%
Diameter (Metric) Mean0.15 μm
Diameter (Metric) Nominal0.15 μm
Diameter (Metric)0.15 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
90 mL
Diameter (Metric)
0.15 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity90 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<10%
Diameter (Metric) Mean0.15 μm
Diameter (Metric) Nominal0.15 μm
Diameter (Metric)0.15 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
15 mL
Diameter (Metric)
0.30 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity15 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean0.30 μm
Diameter (Metric) Nominal0.3 μm
Diameter (Metric)0.30 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
90 mL
Diameter (Metric)
0.30 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity90 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean0.30 μm
Diameter (Metric) Nominal0.3 μm
Diameter (Metric)0.30 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
90 mL
Diameter (Metric)
0.40 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity90 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean0.40 μm
Diameter (Metric) Nominal0.4 μm
Diameter (Metric)0.40 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
15 mL
Diameter (Metric)
0.05 μm
Price (USD)
Full specifications
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity15 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<15%
Diameter (Metric) Mean0.05 μm
Diameter (Metric) Nominal0.05 μm
Diameter (Metric)0.05 μm
Unit SizeEach
Showing 10 of 21Show more
Catalog NumberSpecificationsUnit SizeQuantityDiameter (Metric)Price (USD)
B0200BFull specifications
Each60 mL2 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity60 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean2 μm
Diameter (Metric) Nominal2 μm
Diameter (Metric)2 μm
Unit SizeEach
B0100Full specifications
Each10 mL1 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity10 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean1 μm
Diameter (Metric) Nominal1 μm
Diameter (Metric)1 μm
Unit SizeEach
B0100BFull specifications
Each60 mL1 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity60 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean1 μm
Diameter (Metric) Nominal1 μm
Diameter (Metric)1 μm
Unit SizeEach
B0200Full specifications
Each10 mL2 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity10 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean2 μm
Diameter (Metric) Nominal2 μm
Diameter (Metric)2 μm
Unit SizeEach
B150Full specifications
Each15 mL0.15 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity15 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<10%
Diameter (Metric) Mean0.15 μm
Diameter (Metric) Nominal0.15 μm
Diameter (Metric)0.15 μm
Unit SizeEach
B150BFull specifications
Each90 mL0.15 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity90 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<10%
Diameter (Metric) Mean0.15 μm
Diameter (Metric) Nominal0.15 μm
Diameter (Metric)0.15 μm
Unit SizeEach
B300Full specifications
Each15 mL0.30 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity15 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean0.30 μm
Diameter (Metric) Nominal0.3 μm
Diameter (Metric)0.30 μm
Unit SizeEach
B300BFull specifications
Each90 mL0.30 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity90 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean0.30 μm
Diameter (Metric) Nominal0.3 μm
Diameter (Metric)0.30 μm
Unit SizeEach
B400BFull specifications
Each90 mL0.40 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity90 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<5%
Diameter (Metric) Mean0.40 μm
Diameter (Metric) Nominal0.4 μm
Diameter (Metric)0.40 μm
Unit SizeEach
B50Full specifications
Each15 mL0.05 μmRequest A Quote
AdditiveTrace amounts of surfactant to inhibit agglomeration and promote stability
Concentration1% solids
ContentsDyed polystyrene microspheres in water
ColorBlue
MaterialPolystyrene
Product TypeFluorescent Particle
Dye TypeFirefli Fluorescent Blue (365,388,412/ 445,445,473)
IncludesPackage Insert Sheet
Particle Density1.05g/cm³
Quantity15 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
Uniformity<15%
Diameter (Metric) Mean0.05 μm
Diameter (Metric) Nominal0.05 μm
Diameter (Metric)0.05 μm
Unit SizeEach
Showing 10 of 21Show more
Fluoro-max fluorescent particles emit bright and distinct colors when illuminated by the light of shorter wavelengths than the emission wavelength. This improves their contrast and visibility relative to background materials. These internally dyed polymer particles utilize the exclusive Firefli™ process to incorporate the dye throughout the polymer matrix to produce bright fluorescent colors, minimize photo bleaching and prevent dye leaching into aqueous media.
  • Made of polystyrene (PS)
  • Density of 1.05 g/cm3 and a refractive index of 1.59 @ 589nm (25°C).
  • Packaged as 1% solids
  • Can be detected with an epifluorescence microscope, confocal microscope, fluorometer, fluorescence spectrophotometer or fluorescence activated cell sorter
  • Can also be detected using mineral light or black light (UV)
Note:

Spectral information is approximate and for reference only. The spectral properties of the dyes are dependent on their concentration and physical environment.The exact excitation and emission maxima may vary depending on the size and composition of the particles.

Recommended for:

  • Filter testing
  • Flow tracing
  • Biological research

Contents & Storage

Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. Product comes with stir bar for continual mixing duriong sampling.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text