Thermo Scientific Chemicals

Lactitol monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: B21016.06
5 g, Each
Thermo Scientific Chemicals

Lactitol monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: B21016.06
5 g, Each
Quantity
Catalog number: B21016.06
also known as B21016-06
Price (USD)
Price: 216.00
Online price: 183.65
Your price:
Quantity
-

Chemical Identifiers

CAS
81025-04-9
IUPAC Name
4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexane-1,2,3,5,6-pentol hydrate
Molecular Formula
C12H26O12
InChI Key
LXMBXZRLTPSWCR-UHFFFAOYNA-N
SMILES
O.OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)CO
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
2.55-7.28% (0.5-1.5 waters)
Optical Rotation
14° ± 1° (c=1 in water)
Melting Point (clear melt)
94.0-101.0°C
Appearance (Color)
White

Description

Lactitol monohydrate is used in low fat foods, anthelmintic, cosmetic and pharmaceutical excipients. It is used in sugar-free candies, cookies, chocolate and ice cream. It is a laxative, which used to treat constipation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lactitol monohydrate is used in low fat foods, anthelmintic, cosmetic and pharmaceutical excipients. It is used in sugar-free candies, cookies, chocolate and ice cream. It is a laxative, which used to treat constipation.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text