Thermo Scientific Chemicals

N-Acetyl-L-phenylalanine, 99%

Catalog number: B23812.06
5 g, Each
Thermo Scientific Chemicals

N-Acetyl-L-phenylalanine, 99%

Catalog number: B23812.06
5 g, Each
Quantity
Catalog number: B23812.06
also known as B23812-06
Price (USD)
Price: 72.20
Online price: 69.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2018-61-3
IUPAC Name
(2S)-2-acetamido-3-phenylpropanoic acid
Molecular Formula
C11H13NO3
InChI Key
CBQJSKKFNMDLON-JTQLQIEISA-N
SMILES
CC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
Appearance (Color)
White
Optical Rotation
39.7 ± 0.3? (C=1 in methanol)
Form
Powder
Melting Point (clear melt)
166.0-172.0?C
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

It is an essential amino acid produced for medical, feed, and nutritional applications such as in the preparation of aspartame. It is applied as an antidepressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an essential amino acid produced for medical, feed, and nutritional applications such as in the preparation of aspartame. It is applied as an antidepressant.

Solubility
Solubility in methanol (almost transparency). Soluble in acetone and ethanol (20 mg/ ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text