Taq DNA Polymerase, native (5 U/μL)
Thermo Scientific™

Taq DNA Polymerase, native (5 U/μL)

Thermo Scientific Native Taq DNA Polymerase is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus.
製品番号(カタログ番号)フォーマット
EP0282Without BSA
EP0072With BSA
製品番号(カタログ番号) EP0282
価格(JPY)
35,700
Each
フォーマット:
Without BSA
一括またはカスタム形式をリクエストする
Thermo Scientific Taq DNA Polymerase, native, is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5' to 3' synthesis of DNA, has no detectable 3' to 5' exonuclease (proofreading) activity, and possesses low 5' to 3' exonuclease activity. In addition, the polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Native Taq DNA Polymerase is preferred for amplification of bacterial DNA sequences homologous to those found in E. coli.

Available with or without the stabilizing agent BSA, Taq DNA Polymerase, native, with BSA, is often considered the best choice when amplifying DNA samples of lower purity, e.g., genomic DNA from mouse tail.

Both native Taq DNA polymerases are supplied with two buffers: 10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at a wide range of magnesium concentrations and decreases unspecific priming

Features

  • Thermostable with half-life more than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescent-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • DNA labeling
  • High throughput PCR

Notes

  • The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
  • The 10X Taq Buffer without Detergent is recommended for microarray experiments.
研究用途にのみご使用ください。診断目的には使用できません。
仕様
フィデリティ(vs. Taq)1X
フォーマットWithout BSA
ホットスタート不可
オーバーハング3'-A
ポリメラーゼTaq DNAポリメラーゼ
製品タイプDNAポリメラーゼ
数量500ユニット
反応形態別のコンポーネント
原料DNA
容量2 mL
濃度10X
使用対象(アプリケーション)Standard PCR
GC-Rich PCR Performance
反応速度スタンダード
Unit SizeEach
組成および保存条件
内容:
• 100 µL Taq DNAポリメラーゼ、天然型(5 U/µL)
• 10X Taqバッファー(KCl)、2×1.25 mL
• 10X Taqバッファー、(NH4)2SO4、2×1.25 mL
• 25 mM MgCl2、2×1.25 mL

-20℃で保存。