Thermo Scientific Chemicals

N-Boc-D-phenylalaninol, 98%, Thermo Scientific Chemicals

Catalog number: H27713.03
1 g, Each
Thermo Scientific Chemicals

N-Boc-D-phenylalaninol, 98%, Thermo Scientific Chemicals

Catalog number: H27713.03
1 g, Each
Quantity
Catalog number: H27713.03
also known as H27713-03
Price (USD)
Price: 71.40
Online price: 61.65
Your price:
Quantity
-

Chemical Identifiers

CAS
106454-69-7
IUPAC Name
tert-butyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Molecular Formula
C14H21NO3
InChI Key
LDKDMDVMMCXTMO-GFCCVEGCSA-N
SMILES
CC(C)(C)OC(=O)N[C@@H](CO)CC1=CC=CC=C1
Appearance (Color)
White
Assay (HPLC)
≥97.5%
Form
Powder
Melting Point (clear melt)
95.0-101.0?C
Optical Rotation
28.7 ± 0.1? (c=1 in chloroform)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text