Thermo Scientific Chemicals

Nisoldipine, 98%, Thermo Scientific Chemicals

Catalog number: H37972.03
1 g, Each
Thermo Scientific Chemicals

Nisoldipine, 98%, Thermo Scientific Chemicals

Catalog number: H37972.03
1 g, Each
Quantity
Catalog number: H37972.03
also known as H37972-03
Price (USD)
Price: 123.00
Online price: 107.65
Your price:
Quantity
-

Chemical Identifiers

CAS
63675-72-9
IUPAC Name
3-methyl 5-(2-methylpropyl) 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Formula
C20H24N2O6
InChI Key
VKQFCGNPDRICFG-UHFFFAOYNA-N
SMILES
COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OCC(C)C
Assay from Supplier's CofA
≥97.5%
Appearance (Color)
White to yellow
Form
Crystalline powder
Melting Point (clear melt)
147-155?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text