Thermo Scientific Chemicals

Nalpha,Nepsilon-Di-Boc-L-lysine, 98%, Thermo Scientific Chemicals

Catalog number: H62019.22
100 g, Each
Thermo Scientific Chemicals

Nalpha,Nepsilon-Di-Boc-L-lysine, 98%, Thermo Scientific Chemicals

Catalog number: H62019.22
100 g, Each
Quantity
Catalog number: H62019.22
also known as H62019-22
Price (USD)
Price: 203.00
Online price: 172.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2483-46-7
IUPAC Name
(2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoic acid
Molecular Formula
C16H30N2O6
InChI Key
FBVSXKMMQOZUNU-LDGXTIHJNA-N
SMILES
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Appearance (Color)
White
Proton NMR
Conforms to structure
Form
Solid
Assay from Suppliers CofA
≥96.0% (HPLC)
Optical Rotation
-2.75° to -3.75° (c=5 in methanol)

Description

Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.

Solubility
Soluble in chloroform, ether, ethyl acetate and methanol

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text