Thermo Scientific Chemicals

Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: H62025.06
5 g, Each
Thermo Scientific Chemicals

Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: H62025.06
5 g, Each
Quantity
Catalog number: H62025.06
also known as H62025-06
Price (USD)
Price: 95.90
Online price: 81.65
Your price:
Quantity
-

Chemical Identifiers

CAS
35761-26-3
IUPAC Name
3-amino-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Molecular Formula
C11H14N2O4
InChI Key
FOXRXVSTFGNURG-UHFFFAOYNA-N
SMILES
NCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Assay (unspecified)
>97.5%

Description

Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.

Solubility
Soluble in dilute acid.

Notes
Hygroscopic. Store in cool, well ventilated area. Keep container tightly closed. Store away from water/moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text