Thermo Scientific Chemicals

N-Acetyl-D-alanine, 98%, Thermo Scientific Chemicals

Catalog number: H63450.06
5 g, Each
Thermo Scientific Chemicals

N-Acetyl-D-alanine, 98%, Thermo Scientific Chemicals

Catalog number: H63450.06
5 g, Each
Quantity
Catalog number: H63450.06
also known as H63450-06
Price (USD)
Price: 131.00
Online price: 111.65
Your price:
Quantity
-

Chemical Identifiers

CAS
19436-52-3
Assay (HPLC)
>97.5%

Description

N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-D-alanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s). N-acetyl-D-alanine may be used to study aglycon pocket specific binding on vancomycin.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text