Thermo Scientific Chemicals

D-Tyrosine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63630.14
25 g, Each
Thermo Scientific Chemicals

D-Tyrosine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63630.14
25 g, Each
Quantity
Catalog number: H63630.14
also known as H63630-14
Price (USD)
Price: 203.00
Online price: 175.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3728-20-9
IUPAC Name
methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride
Molecular Formula
C10H14ClNO3
InChI Key
VXYFARNRGZWHTJ-MVRPGTNWNA-N
SMILES
Cl.COC(=O)[C@H](N)CC1=CC=C(O)C=C1
Assay (HPLC)
>97.5%

Description

D-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text