Thermo Scientific Chemicals

N-Fmoc-N-methyl-L-valine, 95%, Thermo Scientific Chemicals

Catalog number: H63671.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-N-methyl-L-valine, 95%, Thermo Scientific Chemicals

Catalog number: H63671.06
5 g, Each
Quantity
Catalog number: H63671.06
also known as H63671-06
Price (USD)
Price: 361.00
Your price:
Quantity
-

Chemical Identifiers

CAS
84000-11-3
IUPAC Name
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-3-methylbutanoic acid
Molecular Formula
C21H23NO4
InChI Key
YCXXXPZNQXXRIG-IBGZPJMESA-N
SMILES
CC(C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Assay (HPLC)
>94.0%

Description

N-Fmoc-N-methyl-L-valine is used as enzyme substrates and reagents, culture media additives, dyes, stains and indicators. It is also involved in the preparation of N-methylated peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-N-methyl-L-valine is used as enzyme substrates and reagents, culture media additives, dyes, stains and indicators. It is also involved in the preparation of N-methylated peptides.

Solubility
Slightly soluble in water and dimethyl formamide.

Notes
Incompatible with strong oxidizing agents, reducing agents, bases and alkali metals. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text