Thermo Scientific Chemicals

O-tert-Butyl-L-serine, 98%, Thermo Scientific Chemicals

Catalog number: H63812.03
1 g, Each
Thermo Scientific Chemicals

O-tert-Butyl-L-serine, 98%, Thermo Scientific Chemicals

Catalog number: H63812.03
1 g, Each
Quantity
Catalog number: H63812.03
also known as H63812-03
Price (USD)
Price: 48.40
Online price: 41.65
Your price:
Quantity
-

Chemical Identifiers

CAS
18822-58-7
IUPAC Name
2-amino-3-(tert-butoxy)propanoic acid
Molecular Formula
C7H15NO3
InChI Key
DDCPKNYKNWXULB-UHFFFAOYNA-N
SMILES
CC(C)(C)OCC(N)C(O)=O
Assay (HPLC)
>97.5%
Optical Rotation
-16.5? ? -1? (C=2.5 in water)

Description

O-tert-Butyl-L-serine tert-butyl ester hydrochloride is a protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides and preparation of amino-phospholipids. O-tert-Butyl-L-serine (Ser(TBU-OH), a serine hydroxyl protected amino acid, is used in SPPS such as the synthesis of tailed cyclic RGD peptides to prevent glutarimide formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-tert-Butyl-L-serine tert-butyl ester hydrochloride is a protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides and preparation of amino-phospholipids. O-tert-Butyl-L-serine (Ser(TBU-OH), a serine hydroxyl protected amino acid, is used in SPPS such as the synthesis of tailed cyclic RGD peptides to prevent glutarimide formation.

Solubility
Soluble in water.

Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text