Thermo Scientific Chemicals

L-Aspartic acid diethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63974.06
5 g, Each
Thermo Scientific Chemicals

L-Aspartic acid diethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63974.06
5 g, Each
Quantity
Catalog number: H63974.06
also known as H63974-06
Price (USD)
Price: 31.00
Your price:
Quantity
-

Chemical Identifiers

CAS
16115-68-7
IUPAC Name
1,4-diethyl 2-aminobutanedioate hydrochloride
Molecular Formula
C8H16ClNO4
InChI Key
AJOXZAAREAYBQR-UHFFFAOYNA-N
SMILES
Cl.CCOC(=O)CC(N)C(=O)OCC
Assay (HPLC)
>97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text